CAS RN 31135-55-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3,3’,5-Triiodo-L-thyronine 4’-O-Sulfate, TRC
CAS: 31135-55-4 Molecular Formula: C15H12I3NO7S Molecular Weight (g/mol): 731.04 Synonym: Triiodothyronine Sulfate,3,5-Diiodo-O-[3-iodo-4-(sulfooxy)phenyl]-L-tyrosine,3-[4-(4-Hydroxy-3-iodophenoxy)-3,5-diiodophenyl]-L-alanine Hydrogen Sulfate,Triiodothyronine Hydrogen Sulfate,Liothyronine Sulfate,T3 Sulfate; IUPAC Name: (2S)-2-amino-3-[3,5-diiodo-4-(3-iodo-4-sulfooxyphenoxy)phenyl]propanoic acid SMILES: C1=CC(=C(C=C1OC2=C(C=C(C=C2I)C[C@@H](C(=O)O)N)I)I)OS(=O)(=O)O