missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Methylphenoxy)piperidine hydrochloride, 98%, Thermo Scientific™

Product Code. 15445698
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15445698 1 g 1g
15435006 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15445698 Supplier Thermo Scientific Alfa Aesar Supplier No. H59396.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1858256-18-4
Molecular Formula C12H18ClNO
Molecular Weight (g/mol) 227.732
MDL Number MFCD21606599
InChI Key VDGDBAJMBJSQDC-UHFFFAOYSA-N
Synonym 3-2-methylphenoxy piperidine hydrochloride, 3-2-methylphenoxy piperidine hcl, 3-o-tolyloxy piperidine hydrochloride
PubChem CID 121235567
IUPAC Name 3-(2-methylphenoxy)piperidine;hydrochloride
SMILES CC1=CC=CC=C1OC2CCCNC2.Cl

Specifications

Quantity 1 g
Formula Weight 227.73
Percent Purity 98%
Chemical Name or Material 3-(2-Methylphenoxy)piperidine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.