accessibility menu, dialog, popup

UserName

CAS RN 1858256-18-4

CH3ONHHCl

CAS RN 1858256-18-4

IUPAC Name: 3-(2-methylphenoxy)piperidine hydrochloride
Synonyms: 3-(2-methylphenoxy)piperidine hydrochloride
Molecular Weight (g/mol): 227.73
Molecular Formula: C12H18ClNO
InChi Key: VDGDBAJMBJSQDC-UHFFFAOYNA-N
SMILES: Cl.CC1=CC=CC=C1OC1CCCNC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results