missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Methoxyphenoxy)propylamine hydrochloride, 97+%, Thermo Scientific™

Product Code. 15489775
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15489775 250 mg 250mg
15479775 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15489775 Supplier Thermo Scientific Alfa Aesar Supplier No. H52396.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1366407-75-1
Molecular Formula C10H16ClNO2
Molecular Weight (g/mol) 217.69
MDL Number MFCD05053657,MFCD11036189
InChI Key JMXMLVJNABFTOE-UHFFFAOYSA-N
Synonym 3-2-methoxyphenoxy propan-1-amine hydrochloride, 1-3-aminopropoxy-2-methoxybenzene hydrochloride, 3-2-methoxyphenoxy propylamine hydrochloride, 3-2-methoxyphenoxy propan-1-aminehydrochloride, 1-3-aminopropoxy-2-methoxybenzene hcl, 3-2-methoxyphenoxy propylamine hcl, 3-2-methoxyphenoxy ;propan-1-amine hydrochloride, 3-2-methoxyphenoxy propan-1-amine-hydrogen chloride 1/1
PubChem CID 22987842
SMILES Cl.COC1=CC=CC=C1OCCCN

Specifications

Quantity 250 mg
IUPAC Name 3-(2-methoxyphenoxy)propan-1-amine;hydrochloride
Formula Weight 217.7
Percent Purity ≥97%
Chemical Name or Material 3-(2-Methoxyphenoxy)propylamine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.