accessibility menu, dialog, popup

UserName

H3COONH2HCl

CAS RN 1366407-75-1

IUPAC Name: 3-(2-methoxyphenoxy)propan-1-amine hydrochloride
Synonyms: 3-(2-methoxyphenoxy)propan-1-amine hydrochloride
Molecular Weight (g/mol): 217.69
Molecular Formula: C10H16ClNO2
InChi Key: JMXMLVJNABFTOE-UHFFFAOYSA-N
SMILES: Cl.COC1=CC=CC=C1OCCCN
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)