Learn More
3-[[[2-[(Diaminomethylene)amino]thiazol-4-yl]methyl]sulfanyl]propanamide, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
3-[[[2-[(Diaminomethylene)amino]thiazol-4-yl]methyl]sulfanyl]propanamide
Specifications
Specifications
| Chemical Name or Material | 3-[((2-[(Diaminomethylene)-amino]thiazol-4-yl)methyl)sulfanyl]-propanamide |
| CAS | 76824-16-3 |
| Analyte or Component Names | 3-[((2-[(Diaminomethylene)-amino]thiazol-4-yl)methyl)sulfanyl]-propanamide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C8 H13 N5 O S2 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Famotidine Imp. F (EP), Famotidine USP Related Compound F, Famotidine USP RC F, Propanamide, 3-[[[2-[(aminoiminomethyl)amino]-4-thiazolyl]methyl]thio]- (9CI), 3-[[[2-[(Diaminomethylene)-amino]thiazol-4-yl]methyl]sulfanyl]-propanamide, Famotidine Imp. D (EP), Famotidine Related Compound D, Famotidine Impurity D |
| InChI Formula | InChI=1S/C8H13N5OS2/c9-6(14)1-2-15-3-5-4-16-8(12-5)13-7(10)11/h4H,1-3H2,(H2,9,14)(H4,10,11,12,13) |
| SMILES | NC(=Nc1nc(CSCCC(=O)N)cs1)N |
| IUPAC Name | 3-[[2-(diaminomethylideneamino)-1,3-thiazol-4-yl]methylsulfanyl]propanamide |
| Show More |