Learn More
3-[[[2-[(Diaminomethylene)amino]thiazol-4-yl]methyl]sulfanyl]-N-sulfamoylpropanamide Hydrochloride, Mikromol™
Product Code. 17919894
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Product Code. 17919894
Brand: Mikromol MM0029.010025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3-[[[2-[(Diaminomethylene)amino]thiazol-4-yl]methyl]sulfanyl]-N-sulfamoylpropanamide Hydrochloride
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3-[([2-[(Diaminomethylene)amino]thiazol-4-yl]methyl)sulfanyl]-N-sulfamoyl-propanamide Hydrochloride
CAS 76824-17-4
Analyte or Component Names 3-[([2-[(Diaminomethylene)amino]thiazol-4-yl]methyl)sulfanyl]-N-sulfamoyl-propanamide Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C8 H14 N6 O3 S3 . Cl H
Certifications/Compliance ISO 17025
Synonym Famotidine Imp. C (EP), Famotidine USP Related Compound C, Famotidine USP RC C, Propanamide, 3-[[[2-[(aminoiminomethyl)amino]-4-thiazolyl]methyl]thio]-N-(aminosulfonyl)-, hydrochloride (1:1), Propanamide, 3-[[[2-[(aminoiminomethyl)amino]-4-thiazolyl]methyl]thio]-N-(aminosulfonyl)-, monohydrochloride (9CI), 3-[[[2-[(Diaminomethylene)amino]thiazol-4-yl]methyl]sulfanyl]-N-sulfamoyl-propanamide Hydrochloride, Famotidine Imp. C (EP) as Hydrochloride, Famotidine Related Compound C as Hydrochloride, Famotidine Impurity C as Hydrochloride
InChI Formula InChI=1S/C8H14N6O3S3.ClH/c9-7(10)13-8-12-5(4-19-8)3-18-2-1-6(15)14-20(11,16)17;/h4H,1-3H2,(H,14,15)(H2,11,16,17)(H4,9,10,12,13);1H
SMILES Cl.NC(=Nc1nc(CSCCC(=O)NS(=O)(=O)N)cs1)N
IUPAC Name 3-[[2-(diaminomethylideneamino)-1,3-thiazol-4-yl]methylsulfanyl]-N-sulfamoylpropanamide;hydrochloride
Molecular Weight (g/mol) 374.89
Quantity 25 mg
Formula Weight 374.0056 g/mol