Learn More
3-[2-[4-(6-Fluoro-2-benzoxazolyl)-1-piperidinyl]ethyl]-6,7,8,9-tetrahydro-2-methyl-4H-pyrido[1,2-a]pyrimidin-4-one, Mikromol™
Description
3-[2-[4-(6-Fluoro-2-benzoxazolyl)-1-piperidinyl]ethyl]-6,7,8,9-tetrahydro-2-methyl-4H-pyrido[1,2-a]pyrimidin-4-one
Specifications
Specifications
| Chemical Name or Material | 3-[2-[4-(6-Fluoro-2-benzoxazolyl)-1-piperidinyl]ethyl]-6,7,8,9-tetrahydro-2-methyl-4H-pyrido[1,2-a]pyrimidin-4-one |
| CAS | 1005191-81-0 |
| Analyte or Component Names | 3-[2-[4-(6-Fluoro-2-benzoxazolyl)-1-piperidinyl]ethyl]-6,7,8,9-tetrahydro-2-methyl-4H-pyrido[1,2-a]pyrimidin-4-one |
| Shipping Condition | Room Temperature |
| Molecular Formula | C23 H27 F N4 O2 |
| Certifications/Compliance | ISO 17025 |
| Synonym | 4H-Pyrido[1,2-a]pyrimidin-4-one, 3-[2-[4-(6-fluoro-2-benzoxazolyl)-1-piperidinyl]ethyl]-6,7,8,9-tetrahydro-2-methyl-, 3-[2-[4-(6-Fluoro-2-benzoxazolyl)-1-piperidinyl]ethyl]-6,7,8,9-tetrahydro-2-methyl-4H-pyrido[1,2-a]pyrimidin-4-one |
| InChI Formula | InChI=1S/C23H27FN4O2/c1-15-18(23(29)28-10-3-2-4-21(28)25-15)9-13-27-11-7-16(8-12-27)22-26-19-6-5-17(24)14-20(19)30-22/h5-6,14,16H,2-4,7-13H2,1H3 |
| SMILES | CC1=C(CCN2CCC(CC2)c3oc4cc(F)ccc4n3)C(=O)N5CCCCC5=N1 |
| IUPAC Name | 3-[2-[4-(6-fluoro-1,3-benzoxazol-2-yl)piperidin-1-yl]ethyl]-2-methyl-6,7,8,9-tetrahydropyrido[1,2-a]pyrimidin-4-one |
| Show More |