Learn More
(2S)-1-[(2S)-2-[[(1S)-1-Carboxy-3-phenylpropyl]amino]propanoyl]pyrrolidine-2-carboxylic Acid (Enalaprilat), Mikromol™
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
(2S)-1-[(2S)-2-[[(1S)-1-Carboxy-3-phenylpropyl]amino]propanoyl]pyrrolidine-2-carboxylic Acid (Enalaprilat)
Specifications
Specifications
| Chemical Name or Material | Enalaprilat |
| CAS | 76420-72-9 |
| Analyte or Component Names | Enalaprilat |
| Shipping Condition | Room Temperature |
| Molecular Formula | C18 H24 N2 O5 |
| Certifications/Compliance | ISO 17034 |
| Synonym | Enalapril Maleate Imp. C (EP), Enalapril Imp. C (EP), (2S)-1-[(2S)-2-[[(1S)-1-Carboxy-3-phenylpropyl]amino]propanoyl]pyrrolidine-2-carboxylic Acid, Enalaprilat, L-Proline, N-[(1S)-1-carboxy-3-phenylpropyl]-L-alanyl-, L-Proline, 1-[N-(1-carboxy-3-phenylpropyl)-L-alanyl]-, (S)-, N-[(1S)-1-Carboxy-3-phenylpropyl]-L-alanyl-L-proline, Enalapril acid, Enalapril diacid, Enalaprilate, Enalaprilic acid, MK 421 diacid, MK 422, N-[1(S)-Carboxy-3-phenylpropyl]-L-alanyl-L-proline, Enalapril Maleate Impurity C, Enalapril Impurity C |
| InChI Formula | InChI=1S/C18H24N2O5/c1-12(16(21)20-11-5-8-15(20)18(24)25)19-14(17(22)23)10-9-13-6-3-2-4-7-13/h2-4,6-7,12,14-15,19H,5,8-11H2,1H3,(H,22,23)(H,24,25)/t12-,14-,15-/m0/s1 |
| SMILES | C[C@H](N[C@@H](CCc1ccccc1)C(=O)O)C(=O)N2CCC[C@H]2C(=O)O |
| IUPAC Name | (2S)-1-[(2S)-2-[[(1S)-1-carboxy-3-phenylpropyl]amino]propanoyl]pyrrolidine-2-carboxylic acid |
| Show More |