Learn More
(2RS)-2-(Methylamino)-2-phenylbutyl 3,4,5-Trimethoxy-benzoate Hydrochloride (N-Desmethyltrimebutine Hydrochloride), Mikromol™
Description
(2RS)-2-(Methylamino)-2-phenylbutyl 3,4,5-Trimethoxy-benzoate Hydrochloride (N-Desmethyltrimebutine Hydrochloride)
Specifications
Specifications
| Chemical Name or Material | N-Demethyl Trimebutine hydrochloride |
| CAS | 294882-33-0 |
| Analyte or Component Names | N-Demethyl Trimebutine hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H27 N O5 . Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | Trimebutine Imp. E (EP), (2RS)-2-(Methylamino)-2-phenylbutyl 3,4,5-Trimethoxy-benzoate Hydrochloride, Trimebutine Imp. E (EP) as Hydrochloride, N-Desmethyltrimebutine Hydrochloride, Trimebutine Maleate Impurity E as Hydrochloride, Trimebutine Impurity E as Hydrochloride |
| InChI Formula | InChI=1S/C21H27NO5.ClH/c1-6-21(22-2,16-10-8-7-9-11-16)14-27-20(23)15-12-17(24-3)19(26-5)18(13-15)25-4;/h7-13,22H,6,14H2,1-5H3;1H |
| SMILES | Cl.CCC(COC(=O)c1cc(OC)c(OC)c(OC)c1)(NC)c2ccccc2 |
| IUPAC Name | [2-(methylamino)-2-phenylbutyl] 3,4,5-trimethoxybenzoate;hydrochloride |
| Show More |