missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2RS)-2-Cyano-2-phenylbutanamide, Mikromol™
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Description
(2RS)-2-Cyano-2-phenylbutanamide
Specifications
Specifications
| Chemical Name or Material | (2RS)-2-Cyano-2-phenylbutanamide |
| CAS | 80544-75-8 |
| Analyte or Component Names | (2RS)-2-Cyano-2-phenylbutanamide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C11 H12 N2 O |
| Certifications/Compliance | ISO 17025 |
| Synonym | (2RS)-2-Cyano-2-phenylbutanamide, Primidone Imp. D (EP), Primidone Impurity D |
| InChI Formula | InChI=1S/C11H12N2O/c1-2-11(8-12,10(13)14)9-6-4-3-5-7-9/h3-7H,2H2,1H3,(H2,13,14) |
| SMILES | CCC(C#N)(C(=O)N)c1ccccc1 |
| IUPAC Name | 2-cyano-2-phenylbutanamide |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction