missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2E,5S,6E)-7-[4-(4-Fluorophenyl)-2-(N-methylmethanesulfonamido)-6-(propan-2-yl)pyrimidin-5-yl]-5-hydroxyhepta-2,6-dienoic Acid Calcium Salt, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
(2E,5S,6E)-7-[4-(4-Fluorophenyl)-2-(N-methylmethanesulfonamido)-6-(propan-2-yl)pyrimidin-5-yl]-5-hydroxyhepta-2,6-dienoic Acid Calcium Salt
Specifications
Specifications
| Chemical Name or Material | (2E,5S,6E)-7-[4-(4-Fluorophenyl)-2-(N-methylmethanesulfonamido)-6-(propan-2-yl)pyrimidin-5-yl]-5-hydroxyhepta-2,6-dienoic Acid Calcium Salt |
| Analyte or Component Names | (2E,5S,6E)-7-[4-(4-Fluorophenyl)-2-(N-methylmethanesulfonamido)-6-(propan-2-yl)pyrimidin-5-yl]-5-hydroxyhepta-2,6-dienoic Acid Calcium Salt |
| Shipping Condition | Room Temperature |
| Molecular Formula | 2[C22 H25 F N3 O5 S] . Ca |
| Certifications/Compliance | ISO 9001 |
| Synonym | (2E,5S,6E)-7-[4-(4-fluorophenyl)-2-(N-methylmethanesulfonamido)-6-(propan-2-yl)pyrimidin-5-yl]-5-hydroxyhepta-2,6-dienoic acid calcium salt, Rosuvastatin Calcium Imp. N (EP) as Calcium Salt |
| InChI Formula | InChI=1S/2C22H26FN3O5S.Ca/c2*1-14(2)20-18(13-12-17(27)6-5-7-19(28)29)21(15-8-10-16(23)11-9-15)25-22(24-20)26(3)32(4,30)31;/h2*5,7-14,17,27H,6H2,1-4H3,(H,28,29);/q;;+2/p-2/b2*7-5+,13-12+;/t2*17-;/m00./s1 |
| SMILES | [Ca+2].CC(C)c1nc(nc(c1\C=C\[C@@H](O)C\C=C\C(=O)[O-])c2ccc(F)cc2)N(C)S(=O)(=O)C.CC(C)c3nc(nc(c3\C=C\[C@@H](O)C\C=C\C(=O)[O-])c4ccc(F)cc4)N(C)S(=O)(=O)C |
| IUPAC Name | calcium;(2E,5S,6E)-7-[4-(4-fluorophenyl)-2-[methyl(methylsulfonyl)amino]-6-propan-2-ylpyrimidin-5-yl]-5-hydroxyhepta-2,6-dienoate |
| Molecular Weight (g/mol) | 965.107 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction