Learn More
2B4/CD244/SLAMF4 Antibody Pair (HRP), Novus Biologicals™
Product Code. 18629330 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
15 Plates
5 Plates
Unit Size:
Pack of 15
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18629330 5 Plates Pack of 5
18216038 15 Plates Pack of 15
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 18629330 Supplier Novus Biologicals Supplier No. NBP2788455Plates
Rabbit Monoclonal Antibody
2B4/CD244/SLAMF4 Monoclonal specifically detects 2B4/CD244/SLAMF4 in Human samples. It is validated for ELISA, Sandwich ELISA.
TRUSTED_SUSTAINABILITY
Specifications
Antigen 2B4/CD244/SLAMF4
Classification Monoclonal
Conjugate HRP
Dilution Sandwich ELISA
Gene Alias CD244 antigen, CD244 molecule, natural killer cell receptor 2B4, h2B4, NAILNK cell activation-inducing ligand, natural killer cell receptor 2B4, NKR2B4NK cell type I receptor protein 2B4,2B4°CD244 natural killer cell receptor 2B4, Nmrk, SLAMF4NK cell activation inducing ligand NAIL
Gene Symbols CD244
Host Species Rabbit
Quantity 5 Plates
Research Discipline Myeloid derived Suppressor Cell, Phospho Specific
Gene ID (Entrez) 51744
Test Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for 2B4/CD244/SLAMF4.
Content And Storage Storage of components varies. See protocol for specific instructions.