Learn More
2B4/CD244/SLAMF4 Antibody Pair (HRP), Novus Biologicals™
Product Code. 18216038 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
15 Plates
5 Plates
Unit Size:
Pack of 15
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18216038 15 Plates Pack of 15
18629330 5 Plates Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 18216038 Supplier Novus Biologicals Supplier No. NBP27884515PLATES
Rabbit Monoclonal Antibody
2B4/CD244/SLAMF4 Monoclonal specifically detects 2B4/CD244/SLAMF4 in Human samples. It is validated for ELISA, Sandwich ELISA.
TRUSTED_SUSTAINABILITY
Specifications
Antigen 2B4/CD244/SLAMF4
Applications ELISA
Classification Monoclonal
Conjugate HRP
Dilution Sandwich ELISA
Gene Alias CD244 antigen, CD244 molecule, natural killer cell receptor 2B4, h2B4, NAILNK cell activation-inducing ligand, natural killer cell receptor 2B4, NKR2B4NK cell type I receptor protein 2B4,2B4CD244 natural killer cell receptor 2B4, Nmrk, SLAMF4NK cell activation inducing ligand NAIL
Gene Symbols CD244
Host Species Rabbit
Quantity 15 Plates
Research Discipline Myeloid derived Suppressor Cell, Phospho Specific
Gene ID (Entrez) 51744
Test Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for 2B4/CD244/SLAMF4.
Target Species Human
Content And Storage Storage of components varies. See protocol for specific instructions.