missing translation for 'onlineSavingsMsg'
Learn More

2alpha,17alpha-Dimethyl-5alpha-androstane-3beta,17beta-diol, LoGiCal

Product Code. 30462796
Click to view available options
Quantity:
2 mg
Unit Size:
2mg
This item is not returnable. View return policy

Product Code. 30462796

Brand: LoGiCal LGCFOR3331.01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2alpha,17alpha-Dimethyl-5alpha-androstane-3beta,17beta-diol
CAS 80651-64-5
Analyte or Component Names 2alpha,17alpha-Dimethyl-5alpha-androstane-3beta,17beta-diol
Shipping Condition Ice pack (-20°C)
Molecular Formula C21 H36 O2
Certifications/Compliance ISO 9001
Synonym (2α,3β,5α,17β)-2,17-dimethyl-Androstane-3,17-diol, 5α-Androstane-3β,17β-diol, 2α,17-dimethyl- (7CI)
InChI Formula InChI=1S/C21H36O2/c1-13-12-19(2)14(11-18(13)22)5-6-15-16(19)7-9-20(3)17(15)8-10-21(20,4)23/h13-18,22-23H,5-12H2,1-4H3/t13-,14+,15-,16+,17+,18-,19+,20+,21+/m1/s1
Solution Type Neat
SMILES C[C@@H]1C[C@@]2(C)[C@@H](CC[C@@H]3[C@@H]2CC[C@@]4(C)[C@H]3CC[C@]4(C)O)C[C@H]1O
IUPAC Name (2R,3R,5S,8R,9S,10S,13S,14S,17S)-2,10,13,17-tetramethyl-1,2,3,4,5,6,7,8,9,11,12,14,15,16-tetradecahydrocyclopenta[a]phenanthrene-3,17-diol
Industry Type Clinical Testing
Molecular Weight (g/mol) 320.51
Quantity 2 mg
Formula Weight 320.2715 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.