Learn More
Hamilton™ 28 gauge, Raw Tubing, Sold Per Inch
Product Code. 11558813
Click to view available options
Overall Length (English):
Custom Needle Length
Unit Size:
Each
Product Code. 11558813
Brand: Hamilton™ 21028
Only null left
Add to basket
Add to basket
Hamilton offers raw cut, 304 stainless steel tubing by the inch. This tubing is cut to length but the ends are not polished and the tubing is not passivated like a typical finished needle.
Specification Sheet
  • Gauge – 28 gauge
  • Needle Length – Custom Needle Length
  • Autoclavable – Yes
  • Gas Sterilizable – Yes
  • Sales Quantity – Sold Per Inch
  • Hub – No Hub
Specifications
Type No Hub
Diameter (English) Inner 28 G
Overall Length (English) Custom Needle Length
Material Stainless Steel