Learn More
Hamilton™ 28 gauge, Raw Tubing, Sold Per Inch
Click to view available options
Overall Length (English):
Custom Needle Length
Unit Size:
Each
Description
Specification Sheet
- Gauge – 28 gauge
- Needle Length – Custom Needle Length
- Autoclavable – Yes
- Gas Sterilizable – Yes
- Sales Quantity – Sold Per Inch
- Hub – No Hub
Specifications
Specifications
| Type | No Hub |
| Diameter (English) Inner | 28 G |
| Overall Length (English) | Custom Needle Length |
| Material | Stainless Steel |