missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PTP beta/zeta/PTPRZ Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB26881-SP
This item is not returnable.
View return policy
Description
PTP beta/zeta/PTPRZ Monoclonal specifically detects PTP beta/zeta/PTPRZ in Human samples. It is validated for Immunohistochemistry.
Specifications
| PTP beta/zeta/PTPRZ | |
| Monoclonal | |
| LYOPH | |
| Immunohistochemistry 8-25 ug/mL | |
| P23471 | |
| PTPRZ1 | |
| E. coli-derived recombinant human PTP beta/zeta/PTPRZ Gly302-Asn754 Accession # P23471 | |
| 25 μg | |
| Primary | |
| Detects human PTP beta/zeta/PTPRZ in direct ELISAs. | |
| Human | |
| Purified |
| Immunohistochemistry | |
| 279245 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 3F8 CSPG, HPTPZ, phosphacan, protein tyrosine phosphatase, receptor-type, Z polypeptide 1, PTP18, PTPRZ, PTPRZ1, PTPZ, receptor-type, zeta polypeptide 1, RPTPB, RPTPbeta | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5803 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction