Learn More
Sulindac EP Impurity B, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
Description
Sulindac EP Impurity B
Specifications
Specifications
| Chemical Name or Material | Sulindac Sulfone |
| CAS | 59973-80-7 |
| Analyte or Component Names | Sulindac Sulfone |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C20 H17 F O4 S |
| Certifications/Compliance | ISO 17025 |
| Synonym | (Z)-[5-Fluoro-2-methyl-1-[4-(methylsulfonyl)benzylidene]-1H-inden-3-yl]acetic Acid (Sulindac Sulfone), (1Z)-5-Fluoro-2-methyl-1-[[4-(methylsulfonyl)phenyl]methylene]-1H-indene-3-acetic Acid, 1H-Indene-3-acetic acid, 5-fluoro-2-methyl-1-[[4-(methylsulfonyl)phenyl]methylene]-, (Z)-, (1Z)-5-Fluoro-2-methyl-1-[[4-(methylsulfonyl)phenyl]methylene]-1H-indene-3-acetic acid, Aptosyn, Exisulind, FGN 1, Prevatec, Sulindac sulfone, cis-5-Fluoro-2-methyl-1-[p-methylsulfonylbenzylidenyl]indene-3-acetic acid, Sulindac Related Compound B, Sulindac Impurity B |
| InChI Formula | InChI=1S/C20H17FO4S/c1-12-17(9-13-3-6-15(7-4-13)26(2,24)25)16-8-5-14(21)10-19(16)18(12)11-20(22)23/h3-10H,11H2,1-2H3,(H,22,23)/b17-9- |
| SMILES | CC1=C(CC(=O)O)c2cc(F)ccc2/C/1=C\c3ccc(cc3)S(=O)(=O)C |
| IUPAC Name | 2-[(3Z)-6-fluoro-2-methyl-3-[(4-methylsulfonylphenyl)methylidene]inden-1-yl]acetic acid |
| Show More |