missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-threo-Isopropylphenidate Hydrochloride, Mikromol™

Product Code. 17961886
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
This item is not returnable. View return policy

Product Code. 17961886

Brand: Mikromol MM1031.230025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(+/-)-threo-Isopropylphenidate Hydrochloride
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (±)-threo-Isopropylphenidate Hydrochloride
CAS 1262795-94-7
Analyte or Component Names (±)-threo-Isopropylphenidate Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C16 H23 N O2 . Cl H
Certifications/Compliance ISO 9001
Synonym 2-Piperidineacetic acid, α-phenyl-, 1-methylethyl ester, hydrochloride (1:1), (αR,2R)-rel-, (±)-threo-Isopropylphenidate hydrochloride, Dimethylphenidate hydrochloride, Ritalinic acid isopropyl ester hydrochloride
InChI Formula InChI=1S/C16H23NO2.ClH/c1-12(2)19-16(18)15(13-8-4-3-5-9-13)14-10-6-7-11-17-14;/h3-5,8-9,12,14-15,17H,6-7,10-11H2,1-2H3;1H/t14-,15-;/m1./s1
SMILES Cl.CC(C)OC(=O)[C@@H]([C@H]1CCCCN1)c2ccccc2
IUPAC Name propan-2-yl (2R)-2-phenyl-2-[(2R)-piperidin-2-yl]acetate;hydrochloride
Molecular Weight (g/mol) 297.82
Quantity 25 mg
Formula Weight 297.1496 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.