missing translation for 'onlineSavingsMsg'
Learn More

N1,N3-Di[2-[[4-(2-Chlorophenyl)-3-ethoxycarbonyl-5-methoxycarbonyl-6-methyl-1,4-dihydropyridin-2-yl]methoxy]ethyl]benzene-1,3-dicarboxamide, Mikromol™

Product Code. 30051222
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30051222 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30051222 Supplier Mikromol Supplier No. MM0383.250025
 Restricted Item
Estimated Shipment: 21-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

N1,N3-Di[2-[[4-(2-Chlorophenyl)-3-ethoxycarbonyl-5-methoxycarbonyl-6-methyl-1,4-dihydropyridin-2-yl]methoxy]ethyl]benzene-1,3-dicarboxamide
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N1,N3-Di[2-([4-(2-Chlorophenyl)-3-ethoxycarbonyl-5-methoxycarbonyl-6-methyl-1,4-dihydropyridin-2-yl]methoxy)ethyl]benzene-1,3-dicarboxamide
CAS 2309467-38-5
Analyte or Component Names N1,N3-Di[2-([4-(2-Chlorophenyl)-3-ethoxycarbonyl-5-methoxycarbonyl-6-methyl-1,4-dihydropyridin-2-yl]methoxy)ethyl]benzene-1,3-dicarboxamide
Shipping Condition Room Temperature
Molecular Formula C48 H52 Cl2 N4 O12
Certifications/Compliance ISO 9001
Synonym INDEX NAME NOT YET ASSIGNED
InChI Formula InChI=1S/C48H52Cl2N4O12/c1-7-65-47(59)41-35(53-27(3)37(45(57)61-5)39(41)31-16-9-11-18-33(31)49)25-63-22-20-51-43(55)29-14-13-15-30(24-29)44(56)52-21-23-64-26-36-42(48(60)66-8-2)40(32-17-10-12-19-34(32)50)38(28(4)54-36)46(58)62-6/h9-19,24,39-40,53-54H,7-8,20-23,25-26H2,1-6H3,(H,51,55)(H,52,56)
SMILES CCOC(=O)C1=C(COCCNC(=O)c2cccc(c2)C(=O)NCCOCC3=C(C(C(=C(C)N3)C(=O)OC)c4ccccc4Cl)C(=O)OCC)NC(=C(C1c5ccccc5Cl)C(=O)OC)C
IUPAC Name 3-O-ethyl 5-O-methyl 4-(2-chlorophenyl)-2-[2-[[3-[2-[[4-(2-chlorophenyl)-3-ethoxycarbonyl-5-methoxycarbonyl-6-methyl-1,4-dihydropyridin-2-yl]methoxy]ethylcarbamoyl]benzoyl]amino]ethoxymethyl]-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate
Molecular Weight (g/mol) 947.8521
Quantity 25 mg
Formula Weight 946.2959 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.