missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(2-Aminoethyl)-1,2,3,4-tetrahydronaphthalene-1-yl-carboxamide ((1,2,3,4-Tetrahydro-1-naphthyl)formylethylenediamine), Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
N-(2-Aminoethyl)-1,2,3,4-tetrahydronaphthalene-1-yl-carboxamide ((1,2,3,4-Tetrahydro-1-naphthyl)formylethylenediamine)
Specifications
Specifications
| Chemical Name or Material | N-(2-Aminoethyl)-1,2,3,4-tetrahydronaphthalene-1-yl-carboxamide |
| CAS | 84460-89-9 |
| Analyte or Component Names | N-(2-Aminoethyl)-1,2,3,4-tetrahydronaphthalene-1-yl-carboxamide |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C13 H18 N2 O |
| Certifications/Compliance | ISO 9001 |
| Synonym | N-(2-Aminoethyl)-1,2,3,4-tetrahydro-1-naphthalenecarboxamide, 1-Naphthamide, N-(2-aminoethyl)-1,2,3,4-tetrahydro- (6CI) |
| InChI Formula | InChI=1S/C13H18N2O/c14-8-9-15-13(16)12-7-3-5-10-4-1-2-6-11(10)12/h1-2,4,6,12H,3,5,7-9,14H2,(H,15,16) |
| SMILES | NCCNC(=O)C1CCCc2ccccc12 |
| IUPAC Name | N-(2-aminoethyl)-1,2,3,4-tetrahydronaphthalene-1-carboxamide |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction