missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl (2S)-2-Amino-3-(3,4-dihydroxyphenyl)-2-methylpropanoate Hydrochloride (Methyldopa Methyl Ester Hydrochloride), Mikromol™
Description
Methyl (2S)-2-Amino-3-(3,4-dihydroxyphenyl)-2-methylpropanoate Hydrochloride (Methyldopa Methyl Ester Hydrochloride)
Specifications
Specifications
| Chemical Name or Material | Methyl (2S)-2-Amino-3-(3,4-dihydroxyphenyl)-2-methylpropanoate Hydrochloride |
| CAS | 115054-62-1 |
| Analyte or Component Names | Methyl (2S)-2-Amino-3-(3,4-dihydroxyphenyl)-2-methylpropanoate Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C11 H15 N O4 . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | Methyl (2S)-2-Amino-3-(3,4-dihydroxyphenyl)-2-methylpropanoate Hydrochloride, Methyldopa Methyl Ester Hydrochloride, L-Tyrosine, 3-hydroxy-alpha-methyl-, methyl ester, hydrochloride (1:1), L-Tyrosine, 3-hydroxy-alpha-methyl-, methyl ester, hydrochloride (9CI), L-alpha-Methyldopa methyl ester hydrochloride, Carbidopa Impurity B as Hydrochloride |
| InChI Formula | InChI=1S/C11H15NO4.ClH/c1-11(12,10(15)16-2)6-7-3-4-8(13)9(14)5-7;/h3-5,13-14H,6,12H2,1-2H3;1H/t11-;/m0./s1 |
| SMILES | Cl.COC(=O)[C@@](C)(N)Cc1ccc(O)c(O)c1 |
| IUPAC Name | methyl (2S)-2-amino-3-(3,4-dihydroxyphenyl)-2-methylpropanoate;hydrochloride |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?