missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Loratadine EP Impurity A, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
Loratadine EP Impurity A
Specifications
Specifications
| Chemical Name or Material | Ethyl 4-[(11RS)-8-Chloro-11-hydroxy-6,11-dihydro-5H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-yl]piperidine-1-carboxylate |
| CAS | 133284-74-9 |
| Analyte or Component Names | Ethyl 4-[(11RS)-8-Chloro-11-hydroxy-6,11-dihydro-5H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-yl]piperidine-1-carboxylate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C22 H25 Cl N2 O3 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Ethyl 4-[(11RS)-8-Chloro-11-hydroxy-6,11-dihydro-5H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-yl]piperidine-1-carboxylate, Loratadine Imp. A (EP), Loratadine Impurity A |
| InChI Formula | InChI=1S/C22H25ClN2O3/c1-2-28-21(26)25-12-9-17(10-13-25)22(27)19-8-7-18(23)14-16(19)6-5-15-4-3-11-24-20(15)22/h3-4,7-8,11,14,17,27H,2,5-6,9-10,12-13H2,1H3 |
| SMILES | CCOC(=O)N1CCC(CC1)C2(O)c3ccc(Cl)cc3CCc4cccnc24 |
| IUPAC Name | ethyl 4-(8-chloro-11-hydroxy-5,6-dihydrobenzo[1,2]cyclohepta[2,4-b]pyridin-11-yl)piperidine-1-carboxylate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction