missing translation for 'onlineSavingsMsg'
Learn More

Diisopropyl [[(2R)-1-[6-(Benzoylamino-9H-purin-9-yl]propan-2-yloxy]methyl]phosphonate, Mikromol™

Product Code. 30051004
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30051004 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30051004 Supplier Mikromol Supplier No. MM1329.170025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Diisopropyl [[(2R)-1-[6-(Benzoylamino-9H-purin-9-yl]propan-2-yloxy]methyl]phosphonate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diisopropyl [((2R)-1-(6-Benzoylamino-9H-purin-9-yl)propan-2-yloxy)methyl]phosphonate
CAS 627510-49-0
Analyte or Component Names Diisopropyl [((2R)-1-(6-Benzoylamino-9H-purin-9-yl)propan-2-yloxy)methyl]phosphonate
Shipping Condition Room Temperature
Molecular Formula C22 H30 N5 O5 P
Certifications/Compliance ISO 9001
Synonym Phosphonic acid, [[(1R)-2-[6-(benzoylamino)-9H-purin-9-yl]-1-methylethoxy]methyl]-, bis(1-methylethyl) ester (9CI), Diisopropyl [[(2R)-1-[6-(Benzoylamino-9H-purin-9-yl]propan-2-yloxy]methyl]phosphonate, Diisopropyl [[(2R)-1-[6-(Benzoylamino-9H-purin-9-yl]propan-2-yloxy]methyl]phosphonate
InChI Formula InChI=1S/C22H30N5O5P/c1-15(2)31-33(29,32-16(3)4)14-30-17(5)11-27-13-25-19-20(23-12-24-21(19)27)26-22(28)18-9-7-6-8-10-18/h6-10,12-13,15-17H,11,14H2,1-5H3,(H,23,24,26,28)/t17-/m1/s1
SMILES CC(C)OP(=O)(CO[C@H](C)Cn1cnc2c(NC(=O)c3ccccc3)ncnc12)OC(C)C
IUPAC Name N-[9-[(2R)-2-(diisopropoxyphosphorylmethoxy)propyl]purin-6-yl]benzamide
Molecular Weight (g/mol) 475.48
Quantity 25 mg
Formula Weight 475.1985 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.