missing translation for 'onlineSavingsMsg'
Learn More

6-Methylpteridine-2,4-diamine, Mikromol™

Product Code. 17963986
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
This item is not returnable. View return policy

Product Code. 17963986

Brand: Mikromol MM0040.300025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

6-Methylpteridine-2,4-diamine
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 6-Methylpteridine-2,4-diamine
CAS 708-74-7
Analyte or Component Names 6-Methylpteridine-2,4-diamine
Shipping Condition Room Temperature
Molecular Formula C7 H8 N6
Certifications/Compliance ISO 9001
Synonym 2,4-Pteridinediamine, 6-methyl-, Pteridine, 2,4-diamino-6-methyl- (6CI,7CI,8CI), 6-Methyl-2,4-pteridinediamine, 2,4-Diamino-6-methylpteridine, 6-Methyl-2,4-diaminopteridine, NSC 79658, 6-Methylpteridine-2,4-diamine
InChI Formula InChI=1S/C7H8N6/c1-3-2-10-6-4(11-3)5(8)12-7(9)13-6/h2H,1H3,(H4,8,9,10,12,13)
SMILES Cc1cnc2nc(N)nc(N)c2n1
IUPAC Name 6-methylpteridine-2,4-diamine
Molecular Weight (g/mol) 176.179
Quantity 25 mg
Formula Weight 176.081 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.