missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(5RS)-5-Ethyl-2,6-diimino-5-phenyltetrahydropyrimidin-4(1H)-one, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
Description
(5RS)-5-Ethyl-2,6-diimino-5-phenyltetrahydropyrimidin-4(1H)-one
Specifications
Specifications
| Chemical Name or Material | (5RS)-5-ethyl-2,6-diimino-5-phenyltetrahydropyrimidin-4(1H)-one |
| CAS | 69125-70-8 |
| Analyte or Component Names | (5RS)-5-ethyl-2,6-diimino-5-phenyltetrahydropyrimidin-4(1H)-one |
| Shipping Condition | Room Temperature |
| Molecular Formula | C12 H14 N4 O |
| Certifications/Compliance | ISO 17025 |
| Synonym | 2,6-Diamino-5-ethyl-5-phenyl-4(5H)-Pyrimidinone, Phenobarbital Imp. A (EP), Phenobarbital Impurity A |
| InChI Formula | InChI=1S/C12H14N4O/c1-2-12(8-6-4-3-5-7-8)9(13)15-11(14)16-10(12)17/h3-7H,2H2,1H3,(H4,13,14,15,16,17) |
| SMILES | CCC1(C(=N)NC(=N)NC1=O)c2ccccc2 |
| IUPAC Name | 5-ethyl-2,6-diimino-5-phenyl-1,3-diazinan-4-one |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction