missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Amino-4-(2-fluorophenyl)-1-methyl-6-nitroquinolin-2(1H)-one, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
3-Amino-4-(2-fluorophenyl)-1-methyl-6-nitroquinolin-2(1H)-one
Specifications
Specifications
| Chemical Name or Material | 3-Amino-4-(2-fluorophenyl)-1-methyl-6-nitroquinolin-2(1H)-one |
| CAS | 2512213-47-5 |
| Analyte or Component Names | 3-Amino-4-(2-fluorophenyl)-1-methyl-6-nitroquinolin-2(1H)-one |
| Shipping Condition | Room Temperature |
| Molecular Formula | C16H12FN3O3 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 3-Amino-4-(2-fluorophenyl)-1-methyl-6-nitroquinolin-2(1H)-one, Flunitrazepam Impurity C |
| InChI Formula | InChI=1S/C16H12FN3O3/c1-19-13-7-6-9(20(22)23)8-11(13)14(15(18)16(19)21)10-4-2-3-5-12(10)17/h2-8H,18H2,1H3 |
| SMILES | CN1C(=O)C(=C(c2ccccc2F)c3cc(ccc13)[N+](=O)[O-])N |
| IUPAC Name | 3-amino-4-(2-fluorophenyl)-1-methyl-6-nitro-quinolin-2-one |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction