missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2R,3R)-2,3-Dihydroxy-4-[[4-(2-methylpropoxy)phenyl]methylamino]-4-oxo-butanoic Acid, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
(2R,3R)-2,3-Dihydroxy-4-[[4-(2-methylpropoxy)phenyl]methylamino]-4-oxo-butanoic Acid
Specifications
Specifications
| Chemical Name or Material | (2R,3R)-2,3-Dihydroxy-4-[[4-(2-methylpropoxy)phenyl]methylamino]-4-oxo-butanoic Acid |
| Analyte or Component Names | (2R,3R)-2,3-Dihydroxy-4-[[4-(2-methylpropoxy)phenyl]methylamino]-4-oxo-butanoic Acid |
| Shipping Condition | Room Temperature |
| Molecular Formula | C15 H21 N O6 |
| Certifications/Compliance | ISO 17025 |
| Synonym | (2R,3R)-2,3-Dihydroxy-4-[(4-isobutoxyphenyl)methylamino]-4-oxo-butanoic Acid, (2R,3R)-2,3-Dihydroxy-4-[(4-(2-methylpropyloxy)phenyl)methylamino]-4-oxo-butanoic Acid, (2R,3R)-2,3-Dihydroxy-4-[[4-(2-methylpropoxy)phenyl]methylamino]-4-oxo-butanoic Acid |
| InChI Formula | InChI=1S/C15H21NO6/c1-9(2)8-22-11-5-3-10(4-6-11)7-16-14(19)12(17)13(18)15(20)21/h3-6,9,12-13,17-18H,7-8H2,1-2H3,(H,16,19)(H,20,21)/t12-,13-/m1/s1 |
| SMILES | CC(C)COc1ccc(CNC(=O)[C@H](O)[C@@H](O)C(=O)O)cc1 |
| IUPAC Name | (2R,3R)-2,3-dihydroxy-4-[[4-(2-methylpropoxy)phenyl]methylamino]-4-oxobutanoic acid |
| Molecular Weight (g/mol) | 311.33 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction