missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(11bRS)-2-Benzoyl-1,2,3,6,7,11b-hexahydro-4H-pyrazino[2,1-a]isoquinolin-4-one, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
(11bRS)-2-Benzoyl-1,2,3,6,7,11b-hexahydro-4H-pyrazino[2,1-a]isoquinolin-4-one
Specifications
Specifications
| Chemical Name or Material | (11bRS)-2-Benzoyl-1,2,3,6,7,11b-hexahydro-4H-pyrazino(2,1-a)isoquinolin-4-one |
| CAS | 54761-87-4 |
| Analyte or Component Names | (11bRS)-2-Benzoyl-1,2,3,6,7,11b-hexahydro-4H-pyrazino(2,1-a)isoquinolin-4-one |
| Shipping Condition | Room Temperature |
| Molecular Formula | C19 H18 N2 O2 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Praziquantel Imp. A (EP), Praziquantel USP Related Compound A, Praziquantel USP RC A, 4H-Pyrazino[2,1-a]isoquinolin-4-one, 2-benzoyl-1,2,3,6,7,11b-hexahydro-, (11bRS)-2-Benzoyl-1,2,3,6,7,11b-hexahydro-4H-pyrazino[2,1-a]isoquinolin-4-one, Praziquantel Related Compound A, Praziquantel Impurity A |
| InChI Formula | InChI=1S/C19H18N2O2/c22-18-13-20(19(23)15-7-2-1-3-8-15)12-17-16-9-5-4-6-14(16)10-11-21(17)18/h1-9,17H,10-13H2 |
| SMILES | O=C(N1CC2N(CCc3ccccc23)C(=O)C1)c4ccccc4 |
| IUPAC Name | 2-benzoyl-3,6,7,11b-tetrahydro-1H-pyrazino[2,1-a]isoquinolin-4-one |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction