missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-[3-(Dimethylamino)propyl]-1-(4-fluorophenyl)-3-hydroxy-1,3-dihydroisobenzofuran-5-carbonitrile Oxalate, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
1-[3-(Dimethylamino)propyl]-1-(4-fluorophenyl)-3-hydroxy-1,3-dihydroisobenzofuran-5-carbonitrile Oxalate
Specifications
Specifications
| Chemical Name or Material | 1-[3-(Dimethylamino)propyl]-1-(4-fluorophenyl)-3-hydroxy-1,3-dihydroisobenzofuran-5-carbonitrile Oxalate |
| CAS | 1332724-03-4 |
| Analyte or Component Names | 1-[3-(Dimethylamino)propyl]-1-(4-fluorophenyl)-3-hydroxy-1,3-dihydroisobenzofuran-5-carbonitrile Oxalate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H21 F N2 O2 . C2 H2 O4 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 1-[3-(dimethylamino)propyl]-1-(4-fluorophenyl)-3-hydroxy-1,3-dihydroisobenzofuran-5-carbonitrile oxalate, 1-[3-(dimethylamino)propyl]-1-(4-fluorophenyl)-1,3-dihydro-3-hydroxy-5-isobenzofurancarbonitrile ethanedioate (1:1), Citalopram Related Compound B as Oxalate, Citalopram Hydrobromide Impurity B as Oxalate, Escitalopram Oxalate Impurity B as Oxalate, Citalopram Impurity B as Oxalate, Escitalopram Impurity B as Oxalate |
| InChI Formula | InChI=1S/C20H21FN2O2.C2H2O4/c1-23(2)11-3-10-20(15-5-7-16(21)8-6-15)18-9-4-14(13-22)12-17(18)19(24)25-20;3-1(4)2(5)6/h4-9,12,19,24H,3,10-11H2,1-2H3;(H,3,4)(H,5,6) |
| SMILES | CN(C)CCCC1(OC(O)c2cc(ccc12)C#N)c3ccc(F)cc3.OC(=O)C(=O)O |
| IUPAC Name | 1-[3-(dimethylamino)propyl]-1-(4-fluorophenyl)-3-hydroxy-3H-2-benzofuran-5-carbonitrile;oxalic acid |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction