missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tramadol Hydrochloride, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Tramadol Hydrochloride
Specifications
Specifications
| Chemical Name or Material | Tramadol Hydrochloride |
| CAS | 36282-47-0 |
| Analyte or Component Names | Tramadol Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C16 H25 N O2 . Cl H |
| Certifications/Compliance | ISO 17034 |
| Synonym | Tramadol Hydrochloride, (1R,2R)-rel-2-[(Dimethylamino)methyl]-1-(3-methoxyphenyl)cyclohexanol hydrochloride, Adamon, Adolonta, Bellatram, CG 315, Contramal, Crispin, Cyclohexanol, 2-[(dimethylamino)methyl]-1-(3-methoxyphenyl)-, hydrochloride, cis-, Dolana, Mabron, Melanate, NIH 10969, Omnidol, Radol, Tadol, Topalgic, Trabar, Tradol, Tradol-Puren, Tradonal, Tramadex, Tramadol hydrochloride, Tramagetic, Tramagit, Tramal, Tramazac, Tramed, Tramol, Tridol, Ultram, Zumatran, Zydol, cis-Tramadol hydrochloride, cis-(+/-)-2-[(Dimethylamino)methyl]-1-(3-methoxyphenyl)cyclohexanol hydrochloride, (+/-)-trans-2-[(Dimethylamino)methyl]-1-(m-methoxyphenyl)cyclohexanol hydrochloride |
| InChI Formula | InChI=1S/C16H25NO2.ClH/c1-17(2)12-14-7-4-5-10-16(14,18)13-8-6-9-15(11-13)19-3;/h6,8-9,11,14,18H,4-5,7,10,12H2,1-3H3;1H/t14-,16+;/m1./s1 |
| SMILES | Cl.COc1cccc(c1)[C@@]2(O)CCCC[C@@H]2CN(C)C |
| IUPAC Name | (1R,2R)-2-[(dimethylamino)methyl]-1-(3-methoxyphenyl)cyclohexan-1-ol;hydrochloride |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction