missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetryzoline Hydrochloride, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Tetryzoline Hydrochloride
Specifications
Specifications
| Chemical Name or Material | Tetryzoline hydrochloride |
| CAS | 522-48-5 |
| Analyte or Component Names | Tetryzoline hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C13 H16 N2 . Cl H |
| Certifications/Compliance | ISO 17034 |
| Synonym | Tetryzoline Hydrochloride, 4,5-Dihydro-2-(1,2,3,4-tetrahydro-1-naphthalenyl)-1H-imidazole hydrochloride (1:1), 4,5-Dihydro-2-(1,2,3,4-tetrahydro-1-naphthalenyl)-1H-imidazole monohydrochloride, 2-(1,2,3,4-Tetrahydro-1-naphthyl)-2-imidazoline monohydrochloride, 2-(1,2,3,4-Tetrahydro-1-naphthyl)-2-imidazoline hydrochloride, Murine Plus, Soothe, Tetrahydrozoline hydrochloride, Tetryzoline hydrochloride, Tinarhinin, Tizine, Tyzanol, Tyzanol hydrochloride, Tyzine, Tyzine hydrochloride, Visine, Visine hydrochloride, Yxin |
| InChI Formula | InChI=1S/C13H16N2.ClH/c1-2-6-11-10(4-1)5-3-7-12(11)13-14-8-9-15-13;/h1-2,4,6,12H,3,5,7-9H2,(H,14,15);1H |
| SMILES | Cl.C1CC(C2=NCCN2)c3ccccc3C1 |
| IUPAC Name | 2-(1,2,3,4-tetrahydronaphthalen-1-yl)-4,5-dihydro-1H-imidazole;hydrochloride |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction