missing translation for 'onlineSavingsMsg'
Learn More

Testosterone Cypionate, Mikromol™

Product Code. 18093830
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
This item is not returnable. View return policy

Product Code. 18093830

Brand: Mikromol MM3171.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Testosterone Cypionate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Testosterone Cypionate
CAS 58-20-8
Analyte or Component Names Testosterone Cypionate
Shipping Condition Room Temperature
Molecular Formula C27 H40 O3
Certifications/Compliance ISO 17025
Synonym Testosterone, cyclopentanepropionate (6CI,7CI,8CI), Cyclopentanepropionic acid, ester with testosterone (8CI), DepAndro 100, DepAndro 200, Depo, Depo-testosterone, Deposteron, Depotest, Depovirin, NSC 9157, Pertestis, Testex Leo, Testodrin prolongatum, Testosterone cyclopentylpropionate, Testosterone cypionate, Testosterone, 3-cyclopentanepropionate, Virilon, Androst-4-en-3-one, 17-(3-cyclopentyl-1-oxopropoxy)-, (17beta)-, 17beta-Hydroxyandrost-4-en-3-one cyclopentanepropionate, 17beta-Hydroxyandrost-4-en-3-one cyclopentylpropionate, Testosterone 17beta-cyclopentanepropionate, Testosterone 17beta-cyclopentylpropionate, Testosterone 17beta-cypionate
InChI Formula InChI=1S/C27H40O3/c1-26-15-13-20(28)17-19(26)8-9-21-22-10-11-24(27(22,2)16-14-23(21)26)30-25(29)12-7-18-5-3-4-6-18/h17-18,21-24H,3-16H2,1-2H3/t21-,22-,23-,24-,26-,27-/m0/s1
SMILES C[C@]12CC[C@H]3[C@@H](CCC4=CC(=O)CC[C@]34C)[C@@H]1CC[C@@H]2OC(=O)CCC5CCCC5
IUPAC Name [(8R,9S,10R,13S,14S,17S)-10,13-dimethyl-3-oxo-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-17-yl] 3-cyclopentylpropanoate
Molecular Weight (g/mol) 412.6
Quantity 250 mg
Formula Weight 412.2977 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.