missing translation for 'onlineSavingsMsg'
Learn More

Cypermethrin, TRC

Product Code. 18012750
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
Unit Size:
1g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18012750 250 mg 250mg
30138505 1 g 1g
30041224 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18012750 Supplier Toronto Research Chemicals Supplier No. TRCC989000250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52315-07-8
Molecular Formula C22 H19 Cl2 N O3
Molecular Weight (g/mol) 415.07
Synonym Agrometrin, Agrothrin, Almetrin, Ambush C, Ambush CY, Ammo, Ammo (pesticide), Antiborer 3767, Ardap, Arrivo, Assist 25CS, Asymmethrin, Auzar 25 EC, Bandit, Barrage, Barricade, Barricade (insecticide), Barricade 10EC, Basathrin, Battery (insecticide), CCN 52, Challenger, Challenger 25EC, Cilcord, Cipkord, Colt, Colt (pyrethrinoid insecticide), Creokhin, Cybil, Cymbush, Cymet, Cympa-Ti, Cymperator, Cymtop, Cynoff EC, Cynoff WP, Cyper Lacer, Cyper Lacer 5EC, Cyperco, Cyperfor S, Cyperil, Cyperkill, Cypermethrin, Cypermethrin 25EC, Cypor, Demon, Demon TC, Dimcyp, Drago, Dudu Cyper, EXP 5598, Ecofleece Sheep Dip (Non-OP), Ectomin, Ectopor, Excis, FMC 30980, FMC 45497, FMC 45806, FMC 6113, Fenom, Fenom (pesticide), Flytick, Hilcyperin, Holster, JF 5705F, Kinmix, Kordon, Kral, Kreokhin, LW 1, Leptocide, Luseweilei, NRDC 149, NRDC 160, NRDC 166, Neramethrin, Neramethrin EC 50, Nurele, Nurse Green, PYR-VU-TO 2, Peststop B, Peststop B 5SC, Polytrin, Prevail, Prevail (pesticide), Prevail FT, RU 27998, Ralothrin, Ripcord, Ripcord 10, Robust, Ronatak, SF 06646, Sherpa, Sniper, Sniper (pyrethroid insecticide), Summerin, Supercypermethrin, Supercypermethrin forte, Supermethrin, Supersect, Taran, Tianlongbao, Toppel, Trofy, Tsifoks, Ustaad, Vucht 424, WL 43467, WL 8517, YT 305, α-Cyano-m-phenoxybenzyl 3-(2,2-dichlorovinyl)-2,2-dimethylcyclopropanecarboxylate
IUPAC Name [cyano-(3-phenoxyphenyl)methyl] 3-(2,2-dichloroethenyl)-2,2-dimethylcyclopropane-1-carboxylate
SMILES CC1(C)C(C=C(Cl)Cl)C1C(=O)OC(C#N)c2cccc(Oc3ccccc3)c2

Specifications

Analyte or Component Names Cypermethrin
Shipping Condition Room Temperature
InChI Formula InChI=1S/C22H19Cl2NO3/c1-22(2)17(12-19(23)24)20(22)21(26)28-18(13-25)14-7-6-10-16(11-14)27-15-8-4-3-5-9-15/h3-12,17-18,20H,1-2H3
Solution Type Neat
Industry Type Environmental Testing
Quantity 250 mg
Formula Weight 415.0742
Chemical Name or Material Cypermethrin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.