missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amiodarone Hydrochloride, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Amiodarone Hydrochloride
Specifications
Specifications
| Chemical Name or Material | Amiodarone Hydrochloride |
| CAS | 19774-82-4 |
| Analyte or Component Names | Amiodarone Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C25 H29 I2 N O3 . Cl H |
| Certifications/Compliance | ISO 17034 |
| Synonym | Amiodarone Hydrochloride, (2-Butyl-3-benzofuranyl)[4-[2-(diethylamino)ethoxy]-3,5-diiodophenyl]methanone hydrochloride, 2-Butyl-3-benzofuranyl 4-[2-(diethylamino)ethoxy]-3,5-diiodophenyl ketone hydrochloride, Amiodar, Amiodarone hydrochloride, Amiodaronum hydrochloride, Ancoron, Angiodarona, Atlansil, Cordarex, Cordarone, Cordarone X, L 3428, L 3428-Labaz, Miocard, Miodaron, Ortacrone, Pacerone, Ritmocardyl, Rythmarone, SKF 33134A, Trangorex |
| InChI Formula | InChI=1S/C25H29I2NO3.ClH/c1-4-7-11-22-23(18-10-8-9-12-21(18)31-22)24(29)17-15-19(26)25(20(27)16-17)30-14-13-28(5-2)6-3;/h8-10,12,15-16H,4-7,11,13-14H2,1-3H3;1H |
| SMILES | Cl.CCCCc1oc2ccccc2c1C(=O)c3cc(I)c(OCCN(CC)CC)c(I)c3 |
| IUPAC Name | (2-butyl-1-benzofuran-3-yl)-[4-[2-(diethylamino)ethoxy]-3,5-diiodophenyl]methanone;hydrochloride |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction