missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse Lysosomal alpha-Glucosidase/GAA, CF
Click to view available options
Quantity:
25 μg
Unit Size:
25µg
Description
Measured by its ability to release glucose from starch. The specific activity is >5000 pmol/min/μg, as measured under the described conditions.
Specifications
Specifications
| Accession Number | P70699.2 |
| For Use With (Application) | Enzyme Activity |
| Formulation | Supplied as a 0.2 μm filtered solution in Tris and NaCl. |
| Gene ID (Entrez) | 2548 |
| Molecular Weight (g/mol) | M.W. (Observed): 97-107 kDa, under reducing conditions, M.W. (Theroretical): 99 kDa |
| Quantity | 25 μg |
| Source | Chinese Hamster Ovary cell line, CHO-derived mouse Lysosomal alpha-Glucosidase protein Glu70-Ser953 with a N-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <0.10 EU per 1 μg of the protein by the LAL method. |
| Gene Alias | Acid maltase, Aglucosidase alfa, EC 3.2.1.20, glucosidase, alpha; acid, LYAG, lysosomal alpha-glucosidase |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction