Learn More
Human DAP12 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB52401-SP
This item is not returnable.
View return policy
Description
DAP12 Monoclonal antibody specifically detects DAP12 in Human samples. It is validated for nan
Specifications
| DAP12 | |
| 406209 | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7305 | |
| Human | |
| Purified |
| Monoclonal | |
| Unconjugated | |
| DAP12PLOSL, DNAX-activation protein 12, KARAPPLO-SL, KAR-associated protein, killer activating receptor associated protein, Killer-activating receptor-associated protein, TYRO protein tyrosine kinase binding protein, TYRO protein tyrosine kinase-binding protein | |
| Human Dap12 synthetic peptide, Accession # O43914 | |
| 25 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
| IgG2a |