Learn More
Abcam 20X Wash Buffer, 25 mL
Product Code. 32110438
Change view
Click to view available options
Quantity:
25 mL
Unit Size:
25mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32110438 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 32110438 Supplier Abcam Supplier No. ab25227325mL
 Restricted Item
Estimated Shipment: 09-10-2026
Concentrated buffer solution for use in assay applications.
  • 20X concentration
  • Wash buffer for assay kits
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Product Type Wash Solution
Format Bottle
Quantity 25 mL