missing translation for 'onlineSavingsMsg'
Learn More
Learn More
200.7 Rev. 4.4 - Calibration Standard 1: Ag @ 50; B, Ba, Be, Cd, Ce, Co, Cr, Cu, Mn, Ni, Pb, Sr, V @ 200; As, Se, Tl, Zn @ 500; Ca, K, Mg, P @ 1000 μg/mL in 5% HNO3, VHG™
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
Description
200.7 Rev. 4.4 - Calibration Standard 1: Ag @ 50; B, Ba, Be, Cd, Ce, Co, Cr, Cu, Mn, Ni, Pb, Sr, V @ 200; As, Se, Tl, Zn @ 500; Ca, K, Mg, P @ 1000 μg/mL in 5% HNO3
Specifications
Specifications
| Chemical Name or Material | Silver/Barium/Beryllium/Boron/Cadmium/Cerium/Chromium/Cobalt/Copper/Lead/Manganese/Nickel/Strontium/Vanadium/Arsenic/Selenium/Thallium/Zinc/Calcium/Magnesium/Phosphorus/Potassium |
| CAS | 7440-22-4, 7440-39-3, 7440-41-7, 7440-42-8, 7440-43-9, 7440-45-1 |
| Methods | ICP |
| Analyte or Component Names | Silver/Barium/Beryllium/Boron/Cadmium/Cerium/Chromium/Cobalt/Copper/Lead/Manganese/Nickel/Strontium/Vanadium/Arsenic/Selenium/Thallium/Zinc/Calcium/Magnesium/Phosphorus/Potassium |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Dilute HNO3 |
| Quantity | 100 mL |
| Concentration or Composition (by Analyte or Components) | Silver 50 μg/mL/Barium 200 μg/mL/Beryllium 200 μg/mL/Boron 200 μg/mL/Cadmium 200 μg/mL/Cerium 200 μg/mL/Chromium 200 μg/mL/Cobalt 200 μg/mL/Copper 200 μg/mL/Lead 200 μg/mL/Manganese 200 μg/mL/Nickel 200 μg/mL/Strontium 200 μg/mL/Vanadium 200 μg/mL/Arsenic 500 μg/mL/Selenium 500 μg/mL/Thallium 500 μg/mL/Zinc 500 μg/mL/Calcium 1000 μg/mL/Magnesium 1000 μg/mL/Phosphorus 1000 μg/mL/Potassium 1000 μg/mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction