Learn More
Hamilton™ 20 Gauge Custom PTFE Tubing
Product Code. 11598843
Click to view available options
Overall Length (English):
Custom Length
Unit Size:
Each
Product Code. 11598843
Brand: Hamilton™ 20920
Only null left
Add to basket
Add to basket
PTFE tubing has high chemical resistance, low and high temperature capability, resistance to weathering, low friction, and electrical and thermal insulation.
Specification Sheet
  • Tubing Material – PTFE
  • Gauge – 20 gauge
  • Hub – No Hub
  • Length – Custom Length
  • Sales Quantity – 1 each
Specifications
Type No Hub
Diameter (English) Inner 20 G
Overall Length (English) Custom Length
Material PTFE