missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)benzenesulfonyl chloride, 98%

Product Code. 11438066
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Unit Size:
100g
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11438066

Brand: Thermo Scientific Alfa Aesar A14108.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 776-04-5 | C7H4ClF3O2S | 244.612 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 776-04-5
Molecular Formula C7H4ClF3O2S
Molecular Weight (g/mol) 244.612
MDL Number MFCD00051696
InChI Key ZIZGWNOAHUCACM-UHFFFAOYSA-N
Synonym 2-trifluoromethyl benzenesulfonyl chloride, 2-trifluoromethyl benzene-1-sulfonyl chloride, 2-trifluoromethylbenzenesulphonyl chloride, 2-trifluoromethyl benzenesulphonyl chloride, o-trifluoromethyl benzenesulfonyl chloride, o-trifluoromethylbenzenesulfonyl chloride, 2-trifluoromethyl benzene sulfonyl chloride, benzenesulfonyl chloride, 2-trifluoromethyl, alpha,alpha,alpha-trifluorotoluene-2-sulfonyl chloride
PubChem CID 136616
IUPAC Name 2-(trifluoromethyl)benzenesulfonyl chloride
SMILES C1=CC=C(C(=C1)C(F)(F)F)S(=O)(=O)Cl

Specifications

Melting Point 22°C to 24°C
Boiling Point 133°C to 135°C (14 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
UN Number UN3261
Beilstein 2651854
Formula Weight 244.62
Percent Purity 98%
Chemical Name or Material 2-(Trifluoromethyl)benzenesulfonyl chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.