Learn More
2-(Trifluoromethyl)benzeneboronic acid, 97%
Product Code. 11415777
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. 11415777
Brand: Thermo Scientific Alfa Aesar B24343.06
Request Bulk or Custom Format
CAS: 1423-27-4 | C7H6BF3O2 | 189.928 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1423-27-4
Molecular Formula C7H6BF3O2
Molecular Weight (g/mol) 189.928
MDL Number MFCD00236059
InChI Key JNSBEPKGFVENFS-UHFFFAOYSA-N
Synonym 2-trifluoromethyl phenylboronic acid, 2-trifluoromethylphenylboronic acid, 2-trifluoromethyl phenyl boronic acid, 2-trifluoromethyl benzeneboronic acid, 2-trifluoromethyl phenyl boranediol, o-trifluoromethylphenylboronic acid, 2-boronobenzotrifluoride, alpha,alpha,alpha-trifluoro-o-tolueneboronic acid
PubChem CID 2734387
IUPAC Name [2-(trifluoromethyl)phenyl]boronic acid
SMILES B(C1=CC=CC=C1C(F)(F)F)(O)O
Specifications
Melting Point 108°C to 110°C
Quantity 5 g
Beilstein 2974465
Formula Weight 189.93
Percent Purity 97%
Chemical Name or Material 2-(Trifluoromethyl)benzeneboronic acid