Learn More
2-Sulfoethyl methacrylate 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Product Code. 30320257
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30320257 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30320257 Supplier Dr. Ehrenstorfer Supplier No. DREA17008100AL100
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Sulfoethyl Methacrylate
CAS 10595-80-9
Analyte or Component Names 2-Sulfoethyl Methacrylate
Shipping Condition +20°C
Molecular Formula C6 H10 O5 S
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym Methacrylic acid, 2-sulfoethyl ester (6CI), Methacrylic acid, ester with 2-hydroxyethanesulfonic acid (7CI,8CI), Ethanesulfonic acid, 2-hydroxy-, methacrylate (8CI), 2-(Methacryloyloxy)ethanesulfonic acid, 2-(Methacryloyloxy)ethylsulfonic acid, 2-Methacryloyloxyethyl-1-sulfonic acid, 2-Sulfoethyl methacrylate, Methacrylic acid 2-hydroxyethanesulfonic acid ester, Sulfoethyl methacrylate, 2-Sulfoethyl Methacrylate
InChI Formula InChI=1S/C6H10O5S/c1-5(2)6(7)11-3-4-12(8,9)10/h1,3-4H2,2H3,(H,8,9,10)
SMILES CC(=C)C(=O)OCCS(=O)(=O)O
IUPAC Name 2-(2-methylprop-2-enoyloxy)ethanesulfonic acid
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 194.21
Quantity 1 mL
Formula Weight 194.0249 g/mol