Learn More
2-sec-Butylthiazole, 98+%
Product Code. 11475563
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. 11475563
Brand: Thermo Scientific Alfa Aesar A13483.14
Request Bulk or Custom Format
CAS: 18277-27-5 | C7H11NS | 141.23 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 18277-27-5
Molecular Formula C7H11NS
Molecular Weight (g/mol) 141.23
MDL Number MFCD00051951
InChI Key MHJSWOZJMPIGJQ-UHFFFAOYNA-N
Synonym 2-sec-butylthiazole, 2-sec-butyl thiazole, 2-1-methylpropyl thiazole, 2-1-methylpropyl-thiazole, thiazole, 2-1-methylpropyl, 2-butan-2-yl-1,3-thiazole, thiazole, 2-sec-butyl, fema no. 3372, 2-s-butylthiazole
PubChem CID 519539
IUPAC Name 2-butan-2-yl-1,3-thiazole
SMILES CCC(C)C1=NC=CS1
Specifications
Density 1.007
Boiling Point 174°C to 175°C
Flash Point 58°C (136°F)
Odor Characteristic
Refractive Index 1.4985
Quantity 25 g
UN Number UN1993
Formula Weight 141.24
Percent Purity ≥98%
Chemical Name or Material 2-sec-Butylthiazole