CAS RN 18277-27-5
CAS RN 18277-27-5
IUPAC Name:
2-(butan-2-yl)-1,3-thiazole
Synonyms:
2-(sec-butyl)thiazole
Molecular Weight (g/mol):
141.23
Molecular Formula:
C7H11NS
InChi Key:
MHJSWOZJMPIGJQ-UHFFFAOYNA-N
SMILES:
CCC(C)C1=NC=CS1
1
–
1
of
1
results
1
2-sec-Butylthiazole, 98+%
CAS: 18277-27-5 Molecular Formula: C7H11NS Molecular Weight (g/mol): 141.23 MDL Number: MFCD00051951 InChI Key: MHJSWOZJMPIGJQ-UHFFFAOYNA-N Synonym: 2-sec-butylthiazole,2-sec-butyl thiazole,2-1-methylpropyl thiazole,2-1-methylpropyl-thiazole,thiazole, 2-1-methylpropyl,2-butan-2-yl-1,3-thiazole,thiazole, 2-sec-butyl,fema no. 3372,2-s-butylthiazole PubChem CID: 519539 IUPAC Name: 2-butan-2-yl-1,3-thiazole SMILES: CCC(C)C1=NC=CS1