Learn More
2-[N-Methyl-N-(pyridin-2-yl)amino]ethanol, Mikromol™
Product Code. 30052472
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30052472 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30052472 Supplier Mikromol Supplier No. MM0522.02
 Restricted Item
Estimated Shipment: 16-11-2026
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2-[N-Methyl-N-(pyridin-2-yl)amino]ethanol
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-[N-Methyl-N-(pyridin-2-yl)amino]ethanol
CAS 122321-04-4
Analyte or Component Names 2-[N-Methyl-N-(pyridin-2-yl)amino]ethanol
Shipping Condition Room Temperature
Molecular Formula C8 H12 N2 O
Certifications/Compliance ISO 9001
Synonym 2-[N-Methyl-N-(pyridin-2-yl)amino]ethanol, Ethanol, 2-(methyl-2-pyridinylamino)-, 2-(Methyl-2-pyridylamino)ethanol, 2-[Methyl(2-pyridinyl)amino]ethanol, 2-[N-Methyl-N-(2-pyridyl)amino]ethanol
InChI Formula InChI=1S/C8H12N2O/c1-10(6-7-11)8-4-2-3-5-9-8/h2-5,11H,6-7H2,1H3
SMILES CN(CCO)c1ccccn1
IUPAC Name 2-[methyl(pyridin-2-yl)amino]ethanol
Molecular Weight (g/mol) 152.19
Quantity 100 mg
Formula Weight 152.095 g/mol