missing translation for 'onlineSavingsMsg'
Learn More

2-Methylbenzyl mercaptan, 97%

Product Code. 11412488
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11412488 5 g 5g
11422488 25 g 25g
11432488 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11412488 Supplier Thermo Scientific Alfa Aesar Supplier No. A15816.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

CAS: 7341-24-4 | C8H10S | 138.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7341-24-4
Molecular Formula C8H10S
Molecular Weight (g/mol) 138.23
MDL Number MFCD00022074
InChI Key PJUDFYDAJBQPEA-UHFFFAOYSA-N
Synonym 2-methylphenyl methanethiol, 2-methylbenzyl mercaptan, 2-methylbenzylmercaptan, 2-methylbenzenemethanethiol, o-methylbenzyl mercaptan, o-tolylmethanethiol, o-tolyl-methanethiol, 2-methylphenyl methane-1-thiol
PubChem CID 288028
IUPAC Name (2-methylphenyl)methanethiol
SMILES CC1=CC=CC=C1CS

Specifications

Boiling Point 56°C to 58°C (5 mmHg)
Odor Stench
Refractive Index 1.572
Quantity 5 g
UN Number UN2810
Beilstein 2040956
Sensitivity Air Sensitive
Formula Weight 138.23
Percent Purity 97%
Chemical Name or Material 2-Methylbenzyl mercaptan

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.