CAS RN 7341-24-4
CAS RN 7341-24-4
IUPAC Name:
(2-methylphenyl)methanethiol
Synonyms:
(2-methylphenyl)methanethiol
Molecular Weight (g/mol):
138.23
Molecular Formula:
C8H10S
InChi Key:
PJUDFYDAJBQPEA-UHFFFAOYSA-N
SMILES:
CC1=CC=CC=C1CS
1
–
1
of
1
results
1
2-Methylbenzyl mercaptan, 97%
CAS: 7341-24-4 Molecular Formula: C8H10S Molecular Weight (g/mol): 138.23 MDL Number: MFCD00022074 InChI Key: PJUDFYDAJBQPEA-UHFFFAOYSA-N Synonym: 2-methylphenyl methanethiol,2-methylbenzyl mercaptan,2-methylbenzylmercaptan,2-methylbenzenemethanethiol,o-methylbenzyl mercaptan,o-tolylmethanethiol,o-tolyl-methanethiol,2-methylphenyl methane-1-thiol PubChem CID: 288028 IUPAC Name: (2-methylphenyl)methanethiol SMILES: CC1=CC=CC=C1CS