missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-4-(trifluoromethoxy)benzaldehyde, 97%, Thermo Scientific™

Product Code. 15435837
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15435837 1 g 1g
15445837 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15435837 Supplier Thermo Scientific Alfa Aesar Supplier No. H33050.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2-Methyl-4-(trifluoromethoxy)benzaldehyde is suitable for use in the synthesis of (E)-4-(trifluoromethoxy)benzaldehyde thiosemicarbazone. It is also used in the synthesis of new azo dyes containing of 5(4H)-oxazolone ring, 2-[4-(trifluoromethoxy)phenyl]-1H-benzimidazole and (E)-2-[4-(trifluoromethoxy)benzylidene]indan-1-one.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to air. Store under dry inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 886763-07-1
Molecular Formula C9H7F3O2
Molecular Weight (g/mol) 204.15
MDL Number MFCD07368717
InChI Key JTPTUKVDAYQHMZ-UHFFFAOYSA-N
Synonym 2-methyl-4-trifluoromethoxy benzaldehyde, 2-methyl-4-trifluoromethoxy-benzaldehyde, 2-formyl-5-trifluoromethoxy toluene
PubChem CID 2783304
IUPAC Name 2-methyl-4-(trifluoromethoxy)benzaldehyde
SMILES CC1=C(C=O)C=CC(OC(F)(F)F)=C1

Specifications

Refractive Index 1.469
Quantity 1 g
Sensitivity Air sensitive
Formula Weight 204.15
Percent Purity 97%
Chemical Name or Material 2-Methyl-4-(trifluoromethoxy)benzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.