accessibility menu, dialog, popup

UserName

CH3OFFFO

CAS RN 886763-07-1

IUPAC Name: 2-methyl-4-(trifluoromethoxy)benzaldehyde
Synonyms: 2-methyl-4-(trifluoromethoxy)benzaldehyde
Molecular Weight (g/mol): 204.15
Molecular Formula: C9H7F3O2
InChi Key: JTPTUKVDAYQHMZ-UHFFFAOYSA-N
SMILES: CC1=C(C=O)C=CC(OC(F)(F)F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results